3,4-(Methylenedioxy)thiobenzamide 97%

Supplier: Thermo Scientific
Warning

Synonyms: Benzo[d][1,3]dioxole-5-carbothioamide, 1,3-Benzodioxole-5-carbothioamide

H51835.03
H51835.03EA 161 EUR
H51835.03
3,4-(Methylenedioxy)thiobenzamide 97%
3,4-(Methylenedioxy)thiobenzamide

Purity: 97% CAS No.: 15884-65-8 Molecular Formula: C8H7NO2S Formula Weight: 181.21 MDL No.: MFCD02677739

Formula: C₈H₇NO₂S
MW: 181,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02677739
CAS Number: 15884-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR