2-Bromo-1-(ethoxymethyl)imidazole 97%

Supplier: Thermo Scientific
Warning

H51860.03 H51860.06
H51860.03EA 227 EUR
H51860.03 H51860.06
2-Bromo-1-(ethoxymethyl)imidazole 97%
2-Bromo-1-(ethoxymethyl)imidazole

Purity: 97% CAS No.: 850429-54-8 Molecular Formula: C6H9BrN2O Formula Weight: 205.05 MDL No.: MFCD04038915

Formula: C₆H₉BrN₂O
MW: 205,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038915
CAS Number: 850429-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR