(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid 95%
Supplier: Thermo Scientific
Synonyms:
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, Fmoc-Pip-OH, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-pipecolic Acid, Fmoc-HoPro-OH, 1-Fmoc-L-pipecolic Acid, Fmoc-HomoPro-OH, (S)-1-Fmoc-2-piperidinecarboxylic Acid
H51981.MD
H51981.03
H51981.06
H51981.MDEA
31.8
EUR
H51981.MD
H51981.03
H51981.06
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid 95%
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
Purity: 95%
CAS No.: 86069-86-5
Molecular Formula: C21H21NO4
Formula Weight: 351.40
Melting Point: 153-158°
Beilstein Reference: 4718405
MDL No.: MFCD00235898
Formula:
C₂₁H₂₁NO₄ MW: 351,40 g/mol Melting Pt: 153…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00235898 CAS Number: 86069-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...