trans-4-Benzyl-L-proline hydrochloride 95%

Supplier: Thermo Scientific

H52041.MD H52041.03
H52041.MDEA 268 EUR
H52041.MD H52041.03
trans-4-Benzyl-L-proline hydrochloride 95%
trans-4-Benzyl-L-proline hydrochloride

Purity: 95% Molecular Formula: C12H15NO2.HCl Formula Weight: 241.72 MDL No.: MFCD15143631

Formula: C₁₂H₁₅NO₂·HCl
MW: 241,72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143631
CAS Number: 1279049-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR