Boc-L-β-homoalanine 95%
Supplier: Thermo Scientific
Synonyms:
Boc-β-HoAla-OH, Boc-β-HomoAla-OH
H52066.MD
H52066.03
H52066.MDEA
85.2
EUR
H52066.MD
H52066.03
Boc-L-β-homoalanine 95%
Boc-L-beta-homoalanine
Purity: 95%
CAS No.: 158851-30-0
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Beilstein Reference: 6800577
MDL No.: MFCD00270345
Formula:
C₉H₁₇NO₄ MW: 203,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00270345 CAS Number: 158851-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...