(1R,3S)-3-Aminocyclopentanecarboxylic acid 95%

Supplier: Thermo Scientific
Warning

H52122.03
H52122.03EA 471 EUR
H52122.03
(1R,3S)-3-Aminocyclopentanecarboxylic acid 95%
(1R,3S)-3-Aminocyclopentanecarboxylic acid

Purity: 95% CAS No.: 71830-08-5 Molecular Formula: C6H11NO2 Formula Weight: 129.16 MDL No.: MFCD00211289

Formula: C₆H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00211289
CAS Number: 71830-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR