trans-N-Boc-4-[2-(trifluoromethyl)benzyl]-L-proline 95%
Supplier: Thermo Scientific
H52137.MD
H52137.03
H52137.MDEA
281
EUR
H52137.MD
H52137.03
trans-N-Boc-4-[2-(trifluoromethyl)benzyl]-L-proline 95%
trans-N-Boc-4-[2-(trifluoromethyl)benzyl]-L-proline
Purity: 95%
CAS No.: 957311-13-6
Molecular Formula: C18H22F3NO4
Formula Weight: 373.37
Melting Point: 117-119°
MDL No.: MFCD16658835
Formula:
C₁₈H₂₂F₃NO₄ MW: 373,37 g/mol Melting Pt: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD16658835 CAS Number: 957311-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...