Fmoc-O-(benzylphospho)-L-serine 95%

Supplier: Thermo Scientific

Synonyms: Fmoc-Ser(PO(OBzl)OH)-OH, Fmoc-Ser(HPO₃Bzl)-OH, Fmoc-Ser(HPO3Bzl)-OH

H52168.MD H52168.03 H52168.06
H52168.MDEA 80 EUR
H52168.MD H52168.03 H52168.06
Fmoc-O-(benzylphospho)-L-serine 95%
Fmoc-O-(benzylphospho)-L-serine

Purity: 95% CAS No.: 158171-14-3 Molecular Formula: C25H24NO8P Formula Weight: 497.44 Beilstein Reference: 7060483 MDL No.: MFCD00797869

Formula: C₂₅H₂₄NO₈P
MW: 497,44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00797869
CAS Number: 158171-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR