Pyridine-4-carboxamidoxime-N-oxide 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-Hydroxy-1-oxy-isonicotinamidine

H52205.03 H52205.06
H52205.03EA 0 EUR
H52205.03 H52205.06
Pyridine-4-carboxamidoxime-N-oxide 97%
Pyridine-4-carboxamidoxime-N-oxide

Purity: 97% CAS No.: 66168-50-1 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 MDL No.: MFCD00206512

Formula: C₆H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00206512
CAS Number: 66168-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR