2-(4-Aminophenyl)-4-(4-pyridyl)thiazole 97%
Supplier: Thermo Scientific
|
Warning
|
H52250.MD
H52250.06
H52250.MDEA
0
EUR
H52250.MD
H52250.06
2-(4-Aminophenyl)-4-(4-pyridyl)thiazole 97%
2-(4-Aminophenyl)-4-(4-pyridyl)thiazole
Purity: 97%
Molecular Formula: C14H11N3S
Formula Weight: 253.32
MDL No.: MFCD12114461
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...