Methyl-4-(5-formyl-2-thienyl)benzoate 95%

Supplier: Thermo Scientific

Synonyms: 4-(5-Formyl-2-thienyl)benzoic acid methyl ester, Methyl-4-(5-formyl-2-thienyl)benzenecarboxylate

H52266.06
H52266.06EA 185 EUR
H52266.06
Methyl-4-(5-formyl-2-thienyl)benzoate 95%
Methyl-4-(5-formyl-2-thienyl)benzoate

Purity: 95% Molecular Formula: C13H10O3S Formula Weight: 246.29 MDL No.: MFCD06200909

Formula: C₁₃H₁₀O₃S
MW: 246,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200909
CAS Number: 639816-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR