9-Benzyl-6-chloro-7-deazapurine 95%
Supplier: Thermo Scientific
|
Warning
|
H52271.03
H52271.06
H52271.03EA
0
EUR
H52271.03
H52271.06
9-Benzyl-6-chloro-7-deazapurine 95%
9-Benzyl-6-chloro-7-deazapurine
Purity: 95%
Molecular Formula: C13H10ClN3
Formula Weight: 243.70
MDL No.: MFCD09953011
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...