4-(1H-Pyrazol-1-ylmethyl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific

Synonyms: 1-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzyl]-1H-pyrazole, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)-1H-pyrazole

H52281.03 H52281.06
H52281.03EA 0 EUR
H52281.03 H52281.06
4-(1H-Pyrazol-1-ylmethyl)benzeneboronic acid pinacol ester 95%
4-(1H-Pyrazol-1-ylmethyl)benzeneboronic acid pinacol ester

Purity: 95% Molecular Formula: C16H21BN2O2 Formula Weight: 284.16 MDL No.: MFCD18827483

Formula: C₁₆H₂₁BN₂O₂
MW: 284,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18827483
CAS Number: 1315281-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR