5'-O-(tert-Butyldimethylsilyl)thymidine ≥97%

Supplier: Thermo Fisher Scientific

H52301.MD H52301.03
H52301.MDEA 0 EUR
H52301.MD H52301.03
5'-O-(tert-Butyldimethylsilyl)thymidine ≥97%
5'-O-(tert-Butyldimethylsilyl)thymidine

Purity: 97+% CAS No.: 40733-28-6 Molecular Formula: C16H28N2O5Si Formula Weight: 356.49 MDL No.: MFCD01631041

Formula: C₁₆H₂₈N₂O₅Si
MW: 356.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631041
CAS Number: 40733-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR