2-Bromo-6-fluorophenylacetonitrile 96%
Supplier: Thermo Scientific
|
Danger
|
H52341.03
H52341.03EA
329
EUR
H52341.03
2-Bromo-6-fluorophenylacetonitrile 96%
2-Bromo-6-fluorophenylacetonitrile
Purity: 96%
CAS No.: 936693-22-0
Molecular Formula: C8H5BrFN
Formula Weight: 214.04
Melting Point: 60-62°
UN No.: UN3439
MDL No.: MFCD11226884
Formula:
C₈H₅BrFN MW: 214,04 g/mol Melting Pt: 60…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD11226884 CAS Number: 936693-22-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...