4-Bromofuran-2-carboxamide 96%
Supplier: Thermo Scientific
|
Warning
|
H52355.MD
H52355.03
H52355.06
H52355.MDEA
61.2
EUR
H52355.MD
H52355.03
H52355.06
4-Bromofuran-2-carboxamide 96%
4-Bromofuran-2-carboxamide
Purity: 96%
CAS No.: 957345-95-8
Molecular Formula: C5H4BrNO
Formula Weight: 189.99
MDL No.: MFCD12755858
Formula:
C₅H₄BrNO MW: 189,99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12755858 CAS Number: 957345-95-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...