2,4,5-Trimethyl-3-pyridinol 97%

Supplier: Thermo Fisher Scientific
Warning

H52357.MD H52357.03
H52357.MDEA 0 EUR
H52357.MD H52357.03
2,4,5-Trimethyl-3-pyridinol 97%
2,4,5-Trimethyl-3-pyridinol

Purity: 97% CAS No.: 5622-78-6 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 172-176° Beilstein Reference: 115543 MDL No.: MFCD17015035

Formula: C₈H₁₁NO
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD17015035
CAS Number: 5622-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR