3-(Dimethylamino)-1-(2-thienyl)-1-propanone hydrochloride ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-[3-(Dimethylamino)propionyl]thiophene Hydrochloride

H52377.MD H52377.03
H52377.MDEA 0 EUR
H52377.MD H52377.03
3-(Dimethylamino)-1-(2-thienyl)-1-propanone hydrochloride ≥97%
3-(Dimethylamino)-1-(2-thienyl)-1-propanone hydrochloride

Purity: 97+% CAS No.: 5424-47-5 Molecular Formula: C9H13NOS.HCl Formula Weight: 219.73 MDL No.: MFCD00126349

Formula: C₉H₁₃NOS
MW: 219,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126349
CAS Number: 5424-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR