3-(2-Methoxyphenoxy)propylamine hydrochloride ≥97%
Supplier: Thermo Scientific
|
Warning
|
H52396.MD
H52396.03
H52396.MDEA
0
EUR
H52396.MD
H52396.03
3-(2-Methoxyphenoxy)propylamine hydrochloride ≥97%
3-(2-Methoxyphenoxy)propylamine hydrochloride
Purity: 97+%
Molecular Formula: C10H15NO2.HCl
Formula Weight: 217.70
MDL No.: MFCD11036189
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...