3-Phenoxypropylamine hydrochloride ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(3-Aminopropoxy)benzene hydrochloride
H52398.MD
H52398.03
H52398.MDEA
0
EUR
H52398.MD
H52398.03
3-Phenoxypropylamine hydrochloride ≥97%
3-Phenoxypropylamine hydrochloride
Purity: 97+%
CAS No.: 83708-39-8
Molecular Formula: C9H13NO.HCl
Formula Weight: 187.67
Beilstein Reference: 3697706
MDL No.: MFCD01325325
Formula:
C₉H₁₃NO·HCl Storage Temperature: Ambient |
MDL Number:
MFCD01325325 CAS Number: 83708-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...