D-erythro-Sphingosine hydrochloride 97%

Supplier: Thermo Fisher Scientific

H52427.MCEA 346 EUR
H52427.MC H52427.ME
D-erythro-Sphingosine hydrochloride 97%
D-erythro-Sphingosine hydrochloride

Purity: 97% CAS No.: 2673-72-5 Molecular Formula: C18H37NO2.HCl Formula Weight: 335.96 MDL No.: MFCD08436977

Formula: C₁₈H₃₇NO₂·HCl
Storage Temperature: Ambient
MDL Number: MFCD08436977
CAS Number: 2673-72-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR