N-Boc-3-dimethylamino-D-alanine 97%
Supplier: Thermo Scientific
H52433.MD
H52433.03
H52433.MDEA
119
EUR
H52433.MD
H52433.03
N-Boc-3-dimethylamino-D-alanine 97%
N-Boc-3-dimethylamino-D-alanine
Purity: 97%
CAS No.: 110755-32-3
Molecular Formula: C10H20N2O4
Formula Weight: 232.28
Formula:
C₁₀H₂₀N₂O₄ MW: 232,28 g/mol Storage Temperature: Ambient |
CAS Number:
110755-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...