4-(trans-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: (E)-(4-(3-ethoxy-3-oxoprop-1-en-1-yl)phenyl)boronic acid

H52552.03 H52552.06
H52552.03EA 121 EUR
H52552.03 H52552.06
4-(trans-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 96%
4-(trans-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid

Purity: 96% CAS No.: 850568-49-9 Molecular Formula: C11H13BO4 Formula Weight: 220.03 Melting Point: 130-134° MDL No.: MFCD04115651

Formula: C₁₁H₁₃BO₄
MW: 220,03 g/mol
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD04115651
CAS Number: 850568-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR