tert-Butyl-3-iodoazetidine-1-carboxylate 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Tert-butyl 3-iodoazetidine-1-carboxylate, 1-BOC-3-iodoazetidine, tert-Butyl 3-iodoazetidine-1-carboxylate, N-Boc-3-iodoazetidine
H52558.03
H52558.06
H52558.03EA
61.8
EUR
H52558.03
H52558.06
tert-Butyl-3-iodoazetidine-1-carboxylate 97%
tert-Butyl-3-iodoazetidine-1-carboxylate
Purity: 97%
CAS No.: 254454-54-1
Molecular Formula: C8H14INO2
Formula Weight: 283.11
Physical Form: Liquid
Sensitivity: Light Sensitive
Appearance: Colourless to pale yellow
MDL No.: MFCD09752821
Formula:
C₈H₁₄INO₂ MW: 283,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09752821 CAS Number: 254454-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...