trans-N-Boc-4-cyano-L-proline 97%
Supplier: Thermo Scientific
|
Warning
|
H52563.MD
H52563.03
H52563.MDEA
441
EUR
H52563.MD
H52563.03
trans-N-Boc-4-cyano-L-proline 97%
trans-N-Boc-4-cyano-L-proline
Purity: 97%
CAS No.: 273221-94-6
Molecular Formula: C11H16N2O4
Formula Weight: 240.26
MDL No.: MFCD01860670
Formula:
C₁₁H₁₆N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD01860670 CAS Number: 273221-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...