(2R,3R)-2-(Boc-amino)-3-benzyloxy-1-butanol 97%

Supplier: Thermo Scientific

Synonyms: Boc-Threoninol(Bzl)

H52565.MD H52565.06
H52565.MDEA 0 EUR
H52565.MD H52565.06
(2R,3R)-2-(Boc-amino)-3-benzyloxy-1-butanol 97%
(2R,3R)-2-(Boc-amino)-3-benzyloxy-1-butanol

Purity: 97% CAS No.: 133565-43-2 Molecular Formula: C16H25NO4 Formula Weight: 295.38 MDL No.: MFCD00235948

Formula: C₁₆H₂₅NO₄
MW: 295,4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235948
CAS Number: 133565-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR