(R)-3-(Boc-amino)-4-hydroxybutyramide 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H52578.MD
H52578.03
H52578.06
H52578.MDEA
0
EUR
H52578.MD
H52578.03
H52578.06
(R)-3-(Boc-amino)-4-hydroxybutyramide 97%
(R)-3-(Boc-amino)-4-hydroxybutyramide
Purity: 97%
Molecular Formula: C9H18N2O4
Formula Weight: 218.25
MDL No.: MFCD00270250
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...