3-Fluoro-5-hydroxybenzeneboronic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Fluoro-5-hydroxybenzeneboronic acid, 3-Fluoro-5-hydroxyphenylboronic acid, (3-Fluoro-5-hydroxyphenyl)boronic acid

H52619.MD H52619.03
H52619.MDEA 0 EUR
H52619.MD H52619.03
3-Fluoro-5-hydroxybenzeneboronic acid 98%
3-Fluoro-5-hydroxybenzeneboronic acid

Purity: 98% CAS No.: 871329-82-7 Molecular Formula: C6H6BFO3 Formula Weight: 155.92 Melting Point: 54-62° MDL No.: MFCD07363778

Formula: C₆H₆BFO₃
MW: 155,92 g/mol
Melting Pt: 54…62 °C
Storage Temperature: Ambient
MDL Number: MFCD07363778
CAS Number: 871329-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR