trans-N-Boc-4-cyano-L-proline methyl ester 97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-tert-Butyl 2-methyl (2S,4R)-4-cyanopyrrolidine-1,2-dicarboxylate

H52733.MD H52733.03
H52733.MDEA 478 EUR
H52733.MD H52733.03
trans-N-Boc-4-cyano-L-proline methyl ester 97%
trans-N-Boc-4-cyano-L-proline methyl ester

Purity: 97% CAS No.: 194163-91-2 Molecular Formula: C12H18N2O4 Formula Weight: 254.29 MDL No.: MFCD03094915

Formula: C₁₂H₁₈N₂O₄
MW: 254,286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094915
CAS Number: 194163-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR