2-(Trifluoromethyl)pyrimidine-4-carboxamide 97%

Supplier: Thermo Fisher Scientific
Warning

H52803.MD H52803.03
H52803.MDEA 0 EUR
H52803.MD H52803.03
2-(Trifluoromethyl)pyrimidine-4-carboxamide 97%
2-(Trifluoromethyl)pyrimidine-4-carboxamide

Purity: 97% CAS No.: 914348-10-0 Molecular Formula: C6H4F3N3O Formula Weight: 191.11 MDL No.: MFCD05662689

Formula: C₆H₄F₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD05662689
CAS Number: 914348-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR