3-Bromo-5-fluorobenzeneboronic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Bromo-5-fluorophenylboronic acid, (3-Bromo-5-fluorophenyl)boronic acid

H52878.06
H52878.06EA 0 EUR
H52878.06
3-Bromo-5-fluorobenzeneboronic acid 95%
3-Bromo-5-fluorobenzeneboronic acid

Purity: 95% CAS No.: 849062-37-9 Molecular Formula: C6H5BBrFO2 Formula Weight: 218.82 Melting Point: 300° MDL No.: MFCD06411356

Formula: C₆H₅BBrFO₂
MW: 218,82 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD06411356
CAS Number: 849062-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR