3-Methoxycarbonyl-5-nitrobenzeneboronic acid 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, 3-Methoxycarbonyl-5-nitrophenylboronic acid, 3-(Methoxycarbonyl)-5-nitrophenylboronic acid, (3-(Methoxycarbonyl)-5-nitrophenyl)boronic acid
H52935.03
H52935.06
H52935.03EA
0
EUR
H52935.03
H52935.06
3-Methoxycarbonyl-5-nitrobenzeneboronic acid 98%
3-Methoxycarbonyl-5-nitrobenzeneboronic acid
Purity: 98%
CAS No.: 117342-20-8
Molecular Formula: C8H8BNO6
Formula Weight: 224.96
Melting Point: 250-255°
MDL No.: MFCD02179469
Formula:
C₈H₈BNO₆ Melting Pt: 250…255 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179469 CAS Number: 117342-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...