4-Hydroxy-2-methylphenylboronic acid ≥98%

Supplier: Thermo Scientific
Warning

H53068.MD H53068.03
H53068.MDEA 0 EUR
H53068.MD H53068.03
4-Hydroxy-2-methylphenylboronic acid ≥98%
4-Hydroxy-2-methylphenylboronic acid

Purity: 98% CAS No.: 493035-82-8 Molecular Formula: C7H9BO3 Formula Weight: 151.96 Melting Point: 100-104° MDL No.: MFCD03788424

Formula: C₇H₉BO₃
MW: 151,96 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD03788424
CAS Number: 493035-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR