(2-Methylsulphinylphenyl)boronic acid 97%

Supplier: Thermo Scientific
Warning

H53209.MD H53209.03
H53209.MDEA 145 EUR
H53209.MD H53209.03
(2-Methylsulphinylphenyl)boronic acid 97%
(2-Methylsulphinylphenyl)boronic acid

Purity: 97% CAS No.: 850567-97-4 Molecular Formula: C7H9BO3S Formula Weight: 184.02 Melting Point: 118-122° MDL No.: MFCD03095261

Formula: C₇H₉BO₃S
MW: 184,02 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD03095261
CAS Number: 850567-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR