1-Phenylvinylboronic acid pinacol ester 95%
Supplier: Thermo Scientific
Synonyms:
4,4,5,5-Tetramethyl-2-(1-phenylvinyl)-1,3,2-dioxaborolane
H53216.03
H53216.03EA
91.1
EUR
H53216.03
1-Phenylvinylboronic acid pinacol ester 95%
1-Phenylvinylboronic acid pinacol ester
Purity: 95%
CAS No.: 143825-84-7
Molecular Formula: C14H19BO2
Formula Weight: 230.11
Melting Point: 40-42°
MDL No.: MFCD06659923
Formula:
C₁₄H₁₉BO₂ MW: 230,11 g/mol Melting Pt: 40…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659923 CAS Number: 143825-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...