2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester 95%
Supplier: Thermo Scientific
H53309.MD
H53309.03
H53309.MDEA
50.9
EUR
H53309.MD
H53309.03
2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester 95%
2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester
Purity: 95%
CAS No.: 441011-76-3
Molecular Formula: C12H16BBrO2
Formula Weight: 282.97
MDL No.: MFCD04039022
Formula:
C₁₂H₁₆BBrO₂ MW: 282,97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039022 CAS Number: 441011-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...