N-(tert-Butoxycarbonyl)propargylamine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-Boc-propargylamine, 2-Propynylcarbamic Acid tert-Butyl Ester, tert-Butyl 2-Propynylcarbamate

H53406.03 H53406.06 H53406.14
H53406.03EA 39.8 EUR
H53406.03 H53406.06 H53406.14
N-(tert-Butoxycarbonyl)propargylamine 97%
N-(tert-Butoxycarbonyl)propargylamine

Purity: 97% CAS No.: 92136-39-5 Molecular Formula: C8H13NO2 Formula Weight: 155.20 Storage Temperature: 2°C to 8°C Flash Point: 93°(199°F) Melting Point: 40-44° MDL No.: MFCD07367245

Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
Melting Pt: 40…44 °C
Flash Pt: 93 °C (199 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD07367245
CAS Number: 92136-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR