1-Phenyl-1-pentyn-3-ol 97%
Supplier: Thermo Scientific
|
Warning
|
H53416.03
H53416.06
H53416.14
H53416.03EA
30.8
EUR
H53416.03
H53416.06
H53416.14
1-Phenyl-1-pentyn-3-ol 97%
1-Phenyl-1-pentyn-3-ol
Purity: 97%
CAS No.: 27975-78-6
Molecular Formula: C11H12O
Formula Weight: 160.22
Flash Point:
Formula:
C₁₁H₁₂O Density: 1,336 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041589 CAS Number: 27975-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...