Ethyl-4,4,4-trifluorobut-2-ynoate 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Ethyl 4,4,4-trifluorobut-2-ynoate
H53425.03
H53425.06
H53425.03EA
249
EUR
H53425.03
H53425.06
Ethyl-4,4,4-trifluorobut-2-ynoate 97%
Ethyl-4,4,4-trifluorobut-2-ynoate
Purity: 97%
CAS No.: 79424-03-6
Molecular Formula: C6H5F3O2
Formula Weight: 166.10
Flash Point: 6°(43°F)
Refractive Index: 1.350
Boiling Point: 96-98°
Beilstein Reference: 3539414
UN No.: UN3272
MDL No.: MFCD00192167
Formula:
C₆H₅F₃O₂ Boiling Pt: 96…98 °C Density: 1,162 Flash Pt: 6 °C (43 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00192167 CAS Number: 79424-03-6 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...