5-Norbornene-2-endo,3-endo-dimethanol 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-endo,3-endo-dimethanol, 5-endo,6-endo-Bis(hydroxymethyl)-2-norbornene
H53454.03
H53454.06
H53454.03EA
86.8
EUR
H53454.03
H53454.06
5-Norbornene-2-endo,3-endo-dimethanol 98%
5-Norbornene-2-endo,3-endo-dimethanol
Purity: 98%
CAS No.: 699-97-8
Molecular Formula: C9H14O2
Formula Weight: 154.21
Melting Point: 82-86°
Boiling Point: 170-175°/25mm
MDL No.: MFCD00674044
Formula:
C₉H₁₄O₂ MW: 154,21 g/mol Boiling Pt: 170…175 °C (25 mmHg) Melting Pt: 82…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674044 CAS Number: 699-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...