1-Triethylsilyl-4-triethylsilyloxy-1-butyne 97%

Supplier: Thermo Scientific
Warning

H53488.06 H53488.14
H53488.06EA 0 EUR
H53488.06 H53488.14
1-Triethylsilyl-4-triethylsilyloxy-1-butyne 97%
1-Triethylsilyl-4-triethylsilyloxy-1-butyne

Purity: 97% CAS No.: 160194-28-5 Molecular Formula: C16H34OSi2 Formula Weight: 298.62 MDL No.: MFCD00671376

Formula: C₁₆H₃₄OSi₂
MW: 298,62 g/mol
Density: 0,834
Storage Temperature: Ambient
MDL Number: MFCD00671376
CAS Number: 160194-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR