Naphthalene-1,4-dithiocarboxamide 97%

Supplier: Thermo Scientific
Warning

H54278.MD H54278.03
H54278.MDEA 0 EUR
H54278.MD H54278.03
Naphthalene-1,4-dithiocarboxamide 97%
Naphthalene-1,4-dithiocarboxamide

Purity: 97% CAS No.: 1347815-21-7 Molecular Formula: C12H10N2S2 Formula Weight: 246.35 MDL No.: MFCD20265223

Formula: C₁₂H₁₀N₂S₂
MW: 246,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20265223
CAS Number: 1347815-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR