3-Phenylsulphonamidopyridine-5-boronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific
Warning

H54339.MD H54339.03
H54339.MDEA 0 EUR
H54339.MD H54339.03
3-Phenylsulphonamidopyridine-5-boronic acid pinacol ester 96%
3-Phenylsulphonamidopyridine-5-boronic acid pinacol ester

Purity: 96% CAS No.: 1083326-28-6 Molecular Formula: C17H21BN2O4S Formula Weight: 360.24 MDL No.: MFCD13190589

Formula: C₁₇H₂₁BN₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD13190589
CAS Number: 1083326-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR