N'-Hydroxypyrimidine-2-carboximidamide 97%
Supplier: Thermo Scientific
|
Warning
|
H54351.MD
H54351.03
H54351.MDEA
0
EUR
H54351.MD
H54351.03
N'-Hydroxypyrimidine-2-carboximidamide 97%
N'-Hydroxypyrimidine-2-carboximidamide
Purity: 97%
CAS No.: 90993-49-0
Molecular Formula: C5H6N4O
Formula Weight: 138.13
Melting Point: 230-235°
MDL No.: MFCD09751609
Formula:
C₅H₆N₄O MW: 138,13 g/mol Melting Pt: 230…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD12093698 CAS Number: 90993-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...