2-Aminopyrimidine-5-carbaldehyde 97%

Supplier: Thermo Scientific
Warning

H54440.06
H54440.06EA 697 EUR
H54440.06
2-Aminopyrimidine-5-carbaldehyde 97%
2-Aminopyrimidine-5-carbaldehyde

Purity: 97% CAS No.: 120747-84-4 Molecular Formula: C5H5N3O Formula Weight: 123.11 MDL No.: MFCD03701714

Formula: C₅H₅N₃O
MW: 123,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701714
CAS Number: 120747-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR