4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile 97%
Supplier: Thermo Scientific
|
Warning
|
H54478.MD
H54478.03
H54478.MDEA
31.5
EUR
H54478.MD
H54478.03
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile 97%
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
Purity: 97%
CAS No.: 127406-79-5
Molecular Formula: C11H8N2S
Formula Weight: 200.26
MDL No.: MFCD01765038
Formula:
C₁₁H₈N₂S Storage Temperature: Ambient |
MDL Number:
MFCD01765038 CAS Number: 127406-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...