6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
6-Methoxy-2-methyl-pyridine-3-boronic acid pinacol ester
H54712.MD
H54712.03
H54712.MDEA
0
EUR
H54712.MD
H54712.03
6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester 96%
6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester
Purity: 96%
CAS No.: 1080028-73-4
Molecular Formula: C13H20BNO3
Formula Weight: 249.11
MDL No.: MFCD12923398
Formula:
C₁₃H₂₀BNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD12923398 CAS Number: 1080028-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...