2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(2-(4-Methoxyphenyl)thiazol-4-yl)acetic acid
H54713.03
H54713.MD
H54713.03EA
95.7
EUR
H54713.03
H54713.MD
2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid 97%
2-[2-(4-Methoxyphenyl)-1,3-thiazol-4-yl]acetic acid
Purity: 97%
CAS No.: 23353-14-2
Molecular Formula: C12H11NO3S
Formula Weight: 249.28
Beilstein Reference: 960002
MDL No.: MFCD01316979
Formula:
C₁₂H₁₁NO₃S MW: 249,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01316979 CAS Number: 23353-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...