Ethyl (2-amino-4-thiazolyl)acetate 97%

Supplier: Thermo Fisher Scientific
Warning

H54717.03 H54717.06
H54717.03EA 0 EUR
H54717.03 H54717.06
Ethyl (2-amino-4-thiazolyl)acetate 97%
Ethyl (2-amino-4-thiazolyl)acetate

Purity: 97% CAS No.: 53266-94-7 Molecular Formula: C7H10N2O2S Formula Weight: 186.23 MDL No.: MFCD00005330

Formula: C₇H₁₀N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00005330
CAS Number: 53266-94-7
EINECS: 258-452-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR