N-(4-Cyanophenyl)guanidine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(4-Cyanophenyl)guanidine

H54761.MD H54761.03
H54761.MDEA 90.3 EUR
H54761.MD H54761.03
N-(4-Cyanophenyl)guanidine 97%
N-(4-Cyanophenyl)guanidine

Purity: 97% CAS No.: 5637-42-3 Molecular Formula: C8H8N4 Formula Weight: 160.18 MDL No.: MFCD16251323

Formula: C₈H₈N₄
MW: 160,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16251323
CAS Number: 5637-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR